Assay Details
Gene Symbol: Pak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106238.1 | NP_001099708.1 | 4 - 5 | 905 | 100 | |
XM_008759119.1 | XP_008757341.1 | 4 - 5 | 2955 | 100 | ||
GenBank mRNA | BC168940.1 | - | 4 - 5 | 915 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
