Assay Details
Gene Symbol: Pak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106238.1 | NP_001099708.1 | 6 - 7 | 1484 | 56 | |
XM_008759119.1 | XP_008757341.1 | 6 - 7 | 3534 | 56 | ||
GenBank mRNA | BC168940.1 | - | 6 - 7 | 1494 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
