Assay Details
Gene Symbol: Dcaf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107935.1 | NP_001101405.1 | 5 - 6 | 1202 | 134 | |
XM_006238066.2 | XP_006238128.1 | 4 - 5 | 1040 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
