Assay Details
Gene Symbol: Dcaf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107935.1 | NP_001101405.1 | 6 - 7 | 1348 | 68 | |
XM_006238066.2 | XP_006238128.1 | 5 - 6 | 1186 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P039631 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P039636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928629 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168488 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P039636 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P039631 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P039631 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P039636 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P039631 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P039636 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P532320 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44622 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11644 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10868719 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868720 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868721 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868723 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868722 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868724 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868725 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10868718 | - | - |
Rat Genome 230 2.0 Array | 1396235_at | - | - |
Rat Genome 230 2.0 Array | 1396235_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500003733.rn.1 | TC0500000570.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500003734.rn.1 | TC0500000570.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500003735.rn.1 | TC0500000570.rn.1 | RTA TC |
Rat Expression Set 230 | 1396235_PM_at | - | - |
Rat Expression Set 230 | 1396235_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868718 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21446084 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354319 | - | - |
