Assay Details
Gene Symbol: B4galt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053287.1 | NP_445739.1 | 1 - 2 | 632 | 96 | |
GenBank mRNA | AA956551.1 | - | 1 - 2 | 168 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
