Assay Details
Gene Symbol: Rgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238138.2 | XP_006238200.1 | 3 - 4 | 358 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
