Assay Details
Gene Symbol: Decr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057197.2 | NP_476545.2 | 8 - 9 | 924 | 62 | |
GenBank mRNA | BC059120.1 | - | 8 - 9 | 912 | 6920489 | 62 |
D00569.1 | - | 8 - 9 | 897 | 62 | ||
FQ209689.1 | - | 8 - 9 | 913 | 62 | ||
FQ210607.1 | - | 8 - 9 | 905 | 62 | ||
FQ210830.1 | - | 4 - 5 | 501 | 62 | ||
FQ212507.1 | - | 8 - 9 | 902 | 62 | ||
FQ218387.1 | - | 8 - 9 | 902 | 62 | ||
FQ219666.1 | - | 8 - 9 | 902 | 62 | ||
FQ220227.1 | - | 8 - 9 | 905 | 62 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
