Assay Details
Gene Symbol: Galt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013089.2 | NP_001013107.1 | 5 - 6 | 577 | 79 | |
XM_006238033.2 | XP_006238095.1 | 5 - 6 | 501 | 79 | ||
XM_008763615.1 | XP_008761837.1 | 3 - 4 | 465 | 79 | ||
GenBank mRNA | BC098756.1 | - | 5 - 6 | 577 | 7370278 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
