Assay Details
Gene Symbol: Galt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013089.2 | NP_001013107.1 | 8 - 9 | 890 | 86 | |
XM_006238033.2 | XP_006238095.1 | 8 - 9 | 814 | 86 | ||
XM_008763615.1 | XP_008761837.1 | 6 - 7 | 778 | 86 | ||
GenBank mRNA | BC098756.1 | - | 8 - 9 | 890 | 7370278 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
