Assay Details
Gene Symbol: Sh3gl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053935.1 | NP_446387.1 | 3 - 4 | 271 | 89 | |
XM_006238359.2 | XP_006238421.1 | 3 - 4 | 1655 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
