Assay Details
Gene Symbol: Sh3gl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053935.1 | NP_446387.1 | 6 - 7 | 711 | 69 | |
XM_006238359.2 | XP_006238421.1 | 6 - 7 | 2095 | 69 | ||
GenBank mRNA | AF009603.1 | - | 3 - 4 | 313 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
