Assay Details
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133412.1 | NP_596903.1 | 4 - 5 | 1092 | 78 | |
XM_006245576.2 | XP_006245638.1 | 4 - 5 | 1162 | 78 | ||
GenBank mRNA | AF321415.1 | - | 4 - 5 | 1092 | 78 |
Gene Symbol: LOC102554121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245586.2 | XP_006245648.1 | 4 - 5 | 1163 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
