Assay Details
Gene Symbol: Stam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109121.2 | NP_001102591.1 | 10 - 11 | 1127 | 79 | |
GenBank mRNA | BC168933.1 | - | 10 - 11 | 1127 | 79 | |
FQ221736.1 | - | 10 - 11 | 1203 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
