Assay Details
Gene Symbol: Suv39h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108883.1 | NP_001102353.1 | 1 - 2 | 267 | 129 | |
XM_008771877.1 | XP_008770099.1 | 1 - 2 | 397 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
