Assay Details
Gene Symbol: Suv39h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108883.1 | NP_001102353.1 | 5 - 6 | 1364 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
chromatin assembly or disassembly
chromatin remodeling
male meiotic nuclear division
biological_process
peptidyl-lysine methylation
histone lysine methylation
histone H3-K9 dimethylation
histone H3-K9 trimethylation
negative regulation of circadian rhythm
negative regulation of transcription, DNA-templated
histone H3-K9 methylation
cellular response to hypoxia
transcription cis-regulatory region binding
RNA polymerase II transcription regulatory region sequence-specific DNA binding
molecular_function
chromatin binding
methyltransferase activity
zinc ion binding
protein methyltransferase activity
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K9 specific)
negative regulation of transcription by RNA polymerase II
chromatin assembly or disassembly
chromatin remodeling
male meiotic nuclear division
biological_process
peptidyl-lysine methylation
histone lysine methylation
histone H3-K9 dimethylation
histone H3-K9 trimethylation
negative regulation of circadian rhythm
negative regulation of transcription, DNA-templated
histone H3-K9 methylation
cellular response to hypoxia
transcription cis-regulatory region binding
RNA polymerase II transcription regulatory region sequence-specific DNA binding
molecular_function
chromatin binding
methyltransferase activity
zinc ion binding
protein methyltransferase activity
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K9 specific)
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P161509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67366 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P161509 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P161509 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P161509 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P161509 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91300 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63097 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10796406 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796410 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796409 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796408 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796407 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10796405 | - | - |
Rat Genome 230 2.0 Array | 1382830_at | - | - |
Rat Genome 230 2.0 Array | 1382830_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1700007038.rn.1 | TC1700001023.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700007040.rn.1 | TC1700001023.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700007039.rn.1 | TC1700001023.rn.1 | RTA TC |
Rat Expression Set 230 | 1382830_PM_at | - | - |
Rat Expression Set 230 | 1382830_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796405 | - | - |
Rat Genome U34 Set | rc_AI071877_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21615006 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353330 | - | - |
