Assay Details
Gene Symbol: Pdzrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271251.1 | NP_001258180.1 | 3 - 4 | 919 | 63 | |
GenBank mRNA | FM040449.1 | - | 1 - 2 | 118 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
