Assay Details
Gene Symbol: Ttll3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108640.2 | NP_001102110.2 | 5 - 6 | 569 | 55 | |
GenBank mRNA | BC168907.1 | - | 4 - 5 | 427 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
