Assay Details
Gene Symbol: Rheb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013216.1 | NP_037348.1 | 1 - 2 | 177 | 116 | |
GenBank mRNA | BC088857.1 | - | 1 - 2 | 231 | 116 | |
FQ212847.1 | - | 1 - 2 | 199 | 116 | ||
FQ214530.1 | - | 1 - 2 | 216 | 116 | ||
FQ218725.1 | - | 1 - 2 | 199 | 116 | ||
FQ225701.1 | - | 1 - 2 | 181 | 116 | ||
FQ230203.1 | - | 1 - 2 | 181 | 116 | ||
U08227.1 | - | 1 - 2 | 177 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
