Assay Details
Gene Symbol: Aasdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250858.2 | XP_006250920.1 | 14 - 15 | 3295 | 62 | |
XM_008770108.1 | XP_008768330.1 | 10 - 11 | 2455 | 62 | ||
XM_008770110.1 | XP_008768332.1 | 11 - 12 | 2554 | 62 | ||
XM_008770111.1 | XP_008768333.1 | 11 - 12 | 2554 | 62 | ||
XM_008770112.1 | XP_008768334.1 | 6 - 7 | 2685 | 62 | ||
XM_008770113.1 | XP_008768335.1 | 7 - 8 | 1644 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
