Assay Details
Gene Symbol: Aasdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250858.2 | XP_006250920.1 | 5 - 6 | 1194 | 81 | |
XM_008770108.1 | XP_008768330.1 | 2 - 3 | 453 | 81 | ||
XM_008770109.1 | XP_008768331.1 | 2 - 3 | 453 | 81 | ||
XM_008770110.1 | XP_008768332.1 | 2 - 3 | 453 | 81 | ||
XM_008770111.1 | XP_008768333.1 | 2 - 3 | 453 | 81 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
