Assay Details
Gene Symbol: Aasdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250858.2 | XP_006250920.1 | 6 - 7 | 1384 | 74 | |
XM_008770108.1 | XP_008768330.1 | 3 - 4 | 643 | 74 | ||
XM_008770109.1 | XP_008768331.1 | 3 - 4 | 643 | 74 | ||
XM_008770110.1 | XP_008768332.1 | 3 - 4 | 643 | 74 | ||
XM_008770111.1 | XP_008768333.1 | 3 - 4 | 643 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
