Assay Details
Gene Symbol: Ube2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106006.1 | NP_001099476.1 | 4 - 5 | 512 | 95 | |
XM_006250934.2 | XP_006250996.1 | 4 - 5 | 709 | 95 | ||
XM_008770140.1 | XP_008768362.1 | 2 - 3 | 309 | 95 | ||
GenBank mRNA | FQ234076.1 | - | 4 - 5 | 490 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
