Assay Details
Gene Symbol: Gabrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080586.1 | NP_542153.1 | 1 - 2 | 201 | 125 | |
GenBank mRNA | X57514.1 | - | 1 - 2 | 201 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
