Assay Details
Gene Symbol: Tmem165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024802.1 | NP_001019973.1 | 4 - 5 | 998 | 62 | |
GenBank mRNA | BC097478.1 | - | 4 - 5 | 998 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
