Assay Details
Gene Symbol: Il20ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107521.1 | NP_001100991.1 | 6 - 7 | 919 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P064003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11022 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P064003 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P064003 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P064003 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18013 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85391 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10701948 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10701946 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10701947 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10701943 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10701944 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10701945 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10701942 | - | - |
Affymetrix Rat Clariom S Assay | PSR0100000759.rn.1 | TC0100000159.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10701942 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21311989 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361857 | - | - |
