Assay Details
Gene Symbol: Katna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004217.4 | NP_001004217.2 | 1 - 2 | 186 | 125 | |
GenBank mRNA | BC097929.1 | - | 1 - 2 | 186 | 125 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC097929.1, NM_001004217.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
