Assay Details
Gene Symbol: Katna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004217.4 | NP_001004217.2 | 2 - 3 | 359 | 77 | |
GenBank mRNA | AY621629.1 | - | 1 - 2 | 163 | 77 | |
BC097929.1 | - | 2 - 3 | 359 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
