Assay Details
Gene Symbol: Hpse2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135762.1 | NP_001129234.1 | 14 - 15 | 1472 | 110 | |
XM_008760445.1 | XP_008758667.1 | 10 - 11 | 3656 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
