Assay Details
Gene Symbol: Dlat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031025.1 | NP_112287.1 | 9 - 10 | 1257 | 112 | |
GenBank mRNA | BC107440.1 | - | 9 - 10 | 1257 | 112 | |
D00092.1 | - | 6 - 7 | 761 | 112 | ||
D10655.1 | - | 8 - 9 | 974 | 112 | ||
M16075.1 | - | 6 - 7 | 761 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
