Assay Details
Gene Symbol: Cbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242965.2 | XP_006243027.1 | 10 - 11 | 1909 | 68 | |
XM_576396.6 | XP_576396.3 | 11 - 12 | 2041 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
