Assay Details
Gene Symbol: Cbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242965.2 | XP_006243027.1 | 5 - 6 | 964 | 71 | |
XM_576396.6 | XP_576396.3 | 5 - 6 | 964 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
