Assay Details
Gene Symbol: Clmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173154.1 | NP_775177.1 | 1 - 2 | 121 | 63 | |
GenBank mRNA | AF302047.1 | - | 1 - 2 | 121 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF302047.1, NM_173154.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
