Assay Details
Gene Symbol: Clmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173154.1 | NP_775177.1 | 5 - 6 | 777 | 60 | |
GenBank mRNA | AF302047.1 | - | 5 - 6 | 777 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
