Assay Details
Gene Symbol: Bco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127712.1 | NP_001121184.1 | 5 - 6 | 689 | 101 | |
GenBank mRNA | BC133725.1 | - | 5 - 6 | 689 | 101 | |
DQ083174.1 | - | 3 - 4 | 353 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
