Assay Details
Gene Symbol: Oaz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109899.2 | NP_001103369.1 | 2 - 3 | 346 | 95 | |
NM_001301297.1 | NP_001288226.1 | 2 - 3 | 343 | 95 | ||
GenBank mRNA | CB586487.1 | - | 2 - 3 | 345 | 95 | |
FQ211793.1 | - | 2 - 3 | 346 | 95 | ||
FQ214772.1 | - | 2 - 3 | 346 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
