Assay Details
Gene Symbol: Oaz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109899.2 | NP_001103369.1 | 1 - 2 | 188 | 98 | |
GenBank mRNA | CB586487.1 | - | 1 - 2 | 187 | 98 | |
FQ211793.1 | - | 1 - 2 | 188 | 98 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001109899.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
