Assay Details
Gene Symbol: Tbcel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014089.1 | NP_001014111.1 | 3 - 4 | 497 | 67 | |
XM_006242859.2 | XP_006242921.1 | 3 - 4 | 3144 | 67 | ||
XM_008766134.1 | XP_008764356.1 | 4 - 5 | 3107 | 67 | ||
GenBank mRNA | BC087163.1 | - | 3 - 4 | 497 | 7135124 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
