Assay Details
Gene Symbol: Tbcel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014089.1 | NP_001014111.1 | 6 - 7 | 1056 | 61 | |
XM_006242859.2 | XP_006242921.1 | 6 - 7 | 3703 | 61 | ||
XM_008766134.1 | XP_008764356.1 | 7 - 8 | 3666 | 61 | ||
GenBank mRNA | BC087163.1 | - | 6 - 7 | 1056 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
