Assay Details
Gene Symbol: Atg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014250.1 | NP_001014272.1 | 2 - 3 | 464 | 134 | |
GenBank mRNA | AM087012.1 | - | 2 - 3 | 464 | 134 | |
BC083602.1 | - | 2 - 3 | 464 | 7133143 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
