Assay Details
Gene Symbol: Clic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009651.1 | NP_001009651.1 | 5 - 6 | 794 | 81 | |
GenBank mRNA | BC088182.1 | - | 5 - 6 | 794 | 81 | |
FQ227497.1 | - | 5 - 6 | 746 | 81 | ||
FQ231721.1 | - | 5 - 6 | 746 | 81 | ||
FQ234033.1 | - | 5 - 6 | 792 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
