Assay Details
Gene Symbol: Tbrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009344.2 | NP_001009344.2 | 2 - 3 | 303 | 87 | |
GenBank mRNA | BC087142.1 | - | 2 - 3 | 264 | 7127799 | 87 |
CO572447.1 | - | 2 - 3 | 303 | 87 | ||
FQ219023.1 | - | 2 - 3 | 235 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
