Assay Details
Gene Symbol: Ccdc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242814.2 | XP_006242876.1 | 11 - 12 | 2111 | 131 | |
XM_006242815.2 | XP_006242877.2 | 11 - 12 | 2100 | 131 | ||
XM_008766078.1 | XP_008764300.1 | 11 - 12 | 2100 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
