Assay Details
Gene Symbol: Ccdc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242814.2 | XP_006242876.1 | 12 - 13 | 2262 | 96 | |
XM_006242815.2 | XP_006242877.2 | 12 - 13 | 2251 | 96 | ||
XM_008766078.1 | XP_008764300.1 | 12 - 13 | 2251 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
