Assay Details
Gene Symbol: Cers4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107117.1 | NP_001100587.1 | 11 - 12 | 1188 | 89 | |
XM_006248804.2 | XP_006248866.1 | 10 - 11 | 1304 | 89 | ||
XM_006248805.2 | XP_006248867.1 | 10 - 11 | 1206 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P105928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179456 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P105928 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P105928 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P105928 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P456876 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104382 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18125 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10759518 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759516 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759521 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759520 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759522 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759514 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759515 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759517 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759523 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759519 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10759513 | - | - |
Rat Genome 230 2.0 Array | 1389741_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389741_at | - | - |
Affymetrix HT Rat Focus Array | 1389741_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389741_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1200008374.rn.1 | TC1200000962.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200008376.rn.1 | TC1200000962.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200008382.rn.1 | TC1200000962.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200008385.rn.1 | TC1200000962.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200008390.rn.1 | TC1200000962.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200008383.rn.1 | TC1200000962.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200008379.rn.1 | TC1200000962.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200008378.rn.1 | TC1200000962.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200008381.rn.1 | TC1200000962.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200008380.rn.1 | TC1200000962.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200008377.rn.1 | TC1200000962.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200008373.rn.1 | TC1200000962.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1200008375.rn.1 | TC1200000962.rn.1 | RTA TC |
Rat Expression Set 230 | 1389741_at | - | - |
Rat Expression Set 230 | 1389741_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10759513 | - | - |
Rat Genome U34 Set | rc_AI136564_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22187499 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356893 | - | - |
