Assay Details
Gene Symbol: Cers4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107117.1 | NP_001100587.1 | 2 - 3 | 184 | 59 | |
XM_006248804.2 | XP_006248866.1 | 1 - 2 | 300 | 59 | ||
XM_006248805.2 | XP_006248867.1 | 1 - 2 | 202 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107117.1, XM_006248804.2, XM_006248805.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
