Assay Details
Gene Symbol: Cers4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107117.1 | NP_001100587.1 | 3 - 4 | 360 | 73 | |
XM_006248804.2 | XP_006248866.1 | 2 - 3 | 476 | 73 | ||
XM_006248805.2 | XP_006248867.1 | 2 - 3 | 378 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
