Assay Details
Gene Symbol: Irak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106791.2 | NP_001100261.2 | 1 - 2 | 164 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106791.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
