Assay Details
Gene Symbol: Ccdc134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024355.1 | NP_001019526.1 | 1 - 2 | 315 | 71 | |
XM_006242113.2 | XP_006242175.1 | 2 - 3 | 228 | 71 | ||
GenBank mRNA | BC088206.1 | - | 1 - 2 | 315 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
