Assay Details
Gene Symbol: Fzd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130536.1 | NP_001124008.1 | 5 - 6 | 1839 | 57 | |
XM_008765449.1 | XP_008763671.1 | 4 - 5 | 2319 | 57 | ||
XM_008765450.1 | XP_008763672.1 | 4 - 5 | 1902 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
