Assay Details
Gene Symbol: Parvg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130583.1 | NP_001124055.1 | 10 - 11 | 843 | 70 | |
XM_006241411.1 | XP_006241473.1 | 10 - 11 | 800 | 70 | ||
XM_006241412.2 | XP_006241474.1 | 10 - 11 | 1053 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
